Potassium iodide 99.0-101.5%, GenAR® USP, Macron Fine Chemicals™
Supplier: Avantor
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Melting Pt:681 °C
:GHS07
:GHS08
Specification Test Results
- Bulk Pharmaceutical Chemical:99.0 - 101.5 %
- USP - Assay (KI)(U.S.P.):99.0 - 101.5 %
- USP - Identification (Potassium):Passes Test
- USP - Identification (Iodide):Passes Test
- USP - Loss on Drying at 105°C:≤0.50 %
- USP - Iodate (IO₃):≤4 mg/kg
- USP - Thiosulfate and Barium:Passes Test
- USP - Alkalinity:Passes Test
- USP - Nitrate, Nitrite, and Ammonia:Passes Test
- EP/BP - Assay (Dried Basis):99.0 - 100.5 %
- EP/BP - Identificaton A (Iodides):Passes Test
- EP/BP - Identification B (Potassium):Passes Test
- EP/BP - Alkalinity:Passes Test
- EP/BP - Appearance (white, granular):Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Iodates:Passes Test
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Thiosulphates:Passes Test
- EP/BP - Iron (Fe):≤20 ppm
- EP/BP - Loss on Drying:≤1.0 %
- Free Alkali, For Information Only:= 10 ppm
- FCC - Assay (KI)(F.C.C.):99.0 - 101.5 %
- FCC - Identification (Potassium):Passes Test
- FCC - Identification (Iodide):Passes Test
- FCC - Identification B (FCC):Passes Test
- FCC - Lead (Pb):≤4 mg/kg
- FCC - Loss on Drying at 105°C:≤0.50 %
- FCC - Iodate (IO₃):≤4 mg/kg
- FCC - Thiosulfate and Barium:Passes Test
- FCC - Nitrate, Nitrite, and Ammonia:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Cell Culture Solutions.pdf
Product Literature
spec_MACR1115-04 spec_MACR1115-05 spec_MACR1115-65