Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hypochlorite solution 5%, J.T.Baker®

Sodium hypochlorite solution 5%, J.T.Baker®

1 L
Catalog # 3936.1000
Supplier:   Avantor
Sodium hypochlorite solution 5%, J.T.Baker®
Catalog # 3936.1000
Supplier:  Avantor
CAS Number:  7681-52-9
Sodium hypochlorite solution 5%, J.T.Baker®

1 L

CAS Number:  7681-52-9
Price
26,36 €
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:7681-52-9
  • Formula:ClNaO
  • MDL Number:MFCD00011120
  • Molecular Weight:74.44 g/mol
  • UN:3212
  • ADR:5.1,II
Specifications

Specifications

  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.