Order Entry
Germany
ContactUsLinkComponent
Calcium chloride dihydrate 99.0-105.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Calcium chloride dihydrate 99.0-105.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Calcium chloride dihydrate 99.0-105.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (CaCl₂):
    74.0 - 78.0 %
  • Assay (CaCl₂·2H₂O):
    99.0 - 105.0 %
  • Insoluble Matter:
    ≤0.01 %
  • pH of 5% Solution at 25°C:
    4.5 - 8.5
  • Oxidizing Substances (as NO₃):
    ≤0.003 %
  • Sulfate (SO₄):
    ≤0.01 %
  • Ammonium (NH₄):
    ≤0.005 %
  • Barium (Ba):
    ≤0.005 %
  • Magnesium (Mg):
    ≤0.005 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.02 %
  • Strontium (Sr):
    ≤0.1 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information