Order Entry
Germany
ContactUsLinkComponent
 
Orthoboric acid ≥99.5% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
Orthoboric acid ≥99.5% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
    ≥99.5%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (H₃BO₃) (by acidimetry):
    ≥ 99.5 %
  • Insoluble in Methanol:
    ≤ 0.005 %
  • Nonvolatile with Methanol:
    ≤ 0.05 %
  • Chloride (Cl):
    ≤ 0.001 %
  • Phosphate (PO₄):
    ≤ 0.001 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Calcium (Ca):
    ≤ 0.005 %
  • Heavy Metals (as Pb):
    ≤ 0.001 %
  • Trace Impurities - Iron (Fe):
    ≤ 0.001 %