Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetone
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
Expand image
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:
67-64-1
:
Formula:
CH₃COCH₃
MDL Number:
MFCD00008765
:
58.08 g/mol
UN:
1090
ADR:
3,II
Related Information
Documentation
Certificates
Search by Lot Number
Documentation