VWR®, Industrial Wipes, Raspy
Supplier: VWR Collection
Certificates
About this item
These spunlace apertured wipes, made from 51% wood pulp and 49% polyester, are slightly abrasive on one side and soft on the other.
- Extremely strong
- Slightly abrasive side for heavy-duty action
- Resistant to acids, solvents and chlorine
- Recyclable
- Food contact certified
Typical applications include: Food and beverage, automotive, industry, printing, laboratories, healthcare and janitorial tasks.
Basis weight: 75 g/m²
Absorption: 760%