Insect Net
Insect net (without handle) – Grid depth = 680 mm
Catalog # BREU16753
Supplier: BREUKHOVEN
Jump to:
Product Details & Documents
Insect net made of nylon with a sturdy aluminium ring of 290 mm.
Delivery information: Telescopic handle not included.
Specifications
- Description:Insect net (without handle) – Grid depth = 680 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



