ContactUsLinkComponent
Steam sterilisation indicators
Steam sterilisation indicators
Catalog # 489-0100
Supplier:  S▄DPACK MEDICA
undefined
Steam sterilisation indicators
Catalog # 489-0100
Supplier:  S▄DPACK MEDICA
Specifications
  • Ø:
    12,5 mm
  • Description:
    Circular labels - steam sterilisation indicators
  • Pk:
    1 Roll
Specifications
About this item
Steam sterilisation. The labels change colour from pink to dark brown after sterilisation.
Packaging: Roll of 5000 labels.