Safety boots, pull on, Jalfrigg
Supplier: Jallatte
Jump to:
About this item
Metal-free safety boots in natural full-grain leather, highly water and oil resistant.
- Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
- Lightweight, non magnetic, perforated composite toecap (200 J)
- Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Fully lined for use in cold environments.
Colour: Beige
Certifications: EN ISO 20345; S3, CI, SRC
14 Options Available Below
Product Details & Documents
Metal-free safety boots in natural full-grain leather, highly water and oil resistant.
- Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
- Lightweight, non magnetic, perforated composite toecap (200 J)
- Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Fully lined for use in cold environments.
Colour: Beige
Certifications: EN ISO 20345; S3, CI, SRC
Product Family Options
Product Information
- Shoe sizeColourAvailabilityPrice