p-Nitrophenyl phosphate DiTris salt ≥98% (by HPCE), Calbiochem®
Catalog # 487655-5
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Form:Off-white to pale yellow solid
- Purity:≥98% by HPLC
- Contaminants (Free-p-Nitrophenol):≤0,05%
- Storage:2 - 8 °C
Specification Test Results
About this item
:
:461.36 g/mol
CAS Number:68189-42-4
Formula:C₆H₆NO₆P
MDL Number:MFCD00037009