Chemical resistant boot covers, Tychem® 2000 C, model POBA
Catalog # 113-0878
Supplier: DuPont
Specifications
- Garment application:Chemical protection,Laboratory
- Colour:Yellow
- Height:480 mm
- Length:420 mm
- Pk:50
Specifications
About this item
Specially designed for use with Tychem® apparel, these boot covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
- Knee-length boot covers with slip-retardant sole and fixing ties
- Sole is partially stitched: splash-proof, not fully liquid-tight
- Anti-static treatment (EN 1149-5) on inside
- Stitched and over-taped seams with barrier tape for protection and strength
Tychem® 2000 C garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure). Tychem® 2000 C is used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
Shoe size: 45 (11)
Certifications: Type PB[3], EN 14126; Cat. III