ContactUsLinkComponent
Bench protectors, Scienceware®, Labmat™
Bench protectors, Scienceware®, Labmat™
Catalog # SIALL2271-1EA
Supplier:  Merck
Bench protectors, Scienceware®, Labmat™
Catalog # SIALL2271-1EA
Supplier:  Merck
Specifications
  • Packed:
    Roll
  • Colour:
    White
  • Description:
    Bench liner, Labmat™
  • L×W:
    15200×500 mm
  • Pk:
    1
Specifications
About this item
Disposable bench and drawer liners protect surfaces from stains and wear.
  • Highly absorbent upper surface traps up to 420 ml/m² of water, dyes, corrosive chemicals or radioactive solutions
  • Flexible polyethylene film backing is non-skid and prevents soak-through of most solutions
  • Rolls are easily cut with a cutter or scissors while roll is retained in dispenser