Order Entry
Germany
ContactUsLinkComponent
Acrylamide/Bis-Acrylamide, LIQUI-GEL™ solution 40% (19:1), MP Biomedicals
Acrylamide/Bis-Acrylamide, LIQUI-GEL™ solution 40% (19:1), MP Biomedicals
  ICNA04800803
Acrylamide/Bis-Acrylamide, LIQUI-GEL™ solution 40% (19:1), MP Biomedicals
  ICNA04800803

 

  • Description:
    Liqui-Gel (29:1), 40% solution
  • Seq:
    02
  • Pk:
    500 mL

 

 

Liqui-Gel is a 40% (w/v) solution of ultra pure acrylamide and bis-acrylamide in deionised water. The gel is formed by the polymerisation of the acrylamide monomer with a co-monomer often N,N'-Methylene-bis-acrylamide, which acts as a cross-linking agent, in the presence of free radicals or a photochemical initiator.

  • Clear and colourless
  • Used in protein and nucleic acid electrophoresis