ContactUsLinkComponent
Dehydrated culture media, DOB media, MP Biomedicals
Dehydrated culture media, DOB media, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Dehydrated culture media, DOB media, MP Biomedicals
Supplier:  MP Biomedicals
Specifications
  • Storage:
    Store sterile YPD agar at ambient temperature (15...30 °C). Powder medium is hygroscopic, therefore minimise exposure to air to prolong shelf life of media.
Specifications