Hand cleanser dispensers, CleanSafe
Supplier: Bode
Certificates
About this item
High-quality overhead dispensing system with special, collapsible 1000 ml bottles with pre-assembled disposable pump dispenser, eliminating the time-consuming reprocessing of reusable pumps.
- Reliable and user-friendly overhead dispenser system made of plastic or stainless steel
- Available as a manual and touchless version
- Suitable for collapsible, 1000 ml CleanSafe bottles with pre-assembled pump dispenser for single use
- Easy pump and bottle change
- Optimal and safe dosing through product-specific pumps
- Integrated lock (key, plastic variants with removable and permanent key)
- Touchless dispenser with reliable infrared sensor technology, status display (battery voltage) and adjustable stroke setting (one, two or three strokes)
- Quick and easy assembly by screwing or gluing (only suitable for plastic variants, adhesive pads already pre-assembled)
Delivery: Delivery incl. mounting material, mounting/operating instructions, touchless versions have batteries included.