0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for VWR® pHenomenal® Meters
Accessories for VWR® pHenomenal® Meters
:
VWR Collection
Accessories for VWR® pHenomenal® Meters
:
VWR Collection
Manual VWR® Storage Vessel