ContactUsLinkComponent
Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Supplier:  DuPont
Certificates
undefined
Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Supplier:  DuPont
Specifications
  • Catalog No:
  • DUPOTF0290TGY00L2X
  • DUPOTF0290TGY00SM
  • DUPOTF0290TGY0034X
  • 113-0876
  • Model:
  • Tychem® 6000 F
  • Tychem® 6000 F
  • Tychem® 6000 F
  • Tychem® 2000 C
  • Closure type:
  • Back fastening
  • Back fastening
  • Back fastening
  • Back fastening
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Chemical protection, Food
  • Chemical protection, Food
  • Chemical protection, Food
  • Chemical protection, Food
  • Weight:
  • 120 g/m²
  • 120 g/m²
  • 120 g/m²
  • 83 g/m²