Glove liners, HyFlex® 48-105
: Ansell
Knitted nylon gloves with PU coated fingertips.
- Optimum protection for light tasks, high level of comfort
- Good freedom of movement for precise handling of small objects in dry conditions
- Elasticated nylon liner for exceptionally close fit
- Excellent alternative to light leather gloves
Ideal for light duty applications, such as assembling, inspecting and packing small components.
: EN 388:2016 (003XA); Cat. II
- Catalog No:
- 112-3784
- 112-3785
- 112-3782
- 112-3786
- 112-3783
- 112-3787
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- EN norm:
- EN 388:2016 (003XA)
- EN 388:2016 (003XA)
- EN 388:2016 (003XA)
- EN 388:2016 (003XA)
- EN 388:2016 (003XA)
- EN 388:2016 (003XA)