EDTA, 0.5M pH 8.0, Corning®
0.5M EDTA, pH 8.0, Plastic bottle, 100 mL
Supplier: Corning
Jump to:
Product Details & Documents
EDTA (ethylenediaminetetraacetic acid) is a polyamino carboxylic acid. It is a colorless, water-soluble solid and is useful as a chelating agent due to its ability to "sequester" metal ions such as Ca2+ and Fe3+. These cations are responsible for various undesirable effects in molecular biological and cell culture processes. After being bound by EDTA, these metal ions remain in the solution but exhibit diminished reactivity. The use of EDTA therefore allows for greater control over many experiments.
- Complete Certificate of Analysis available for each production lot, along with full formulation information Produced under the highest industry standards to ensre superior results.
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Description:0.5M EDTA, pH 8.0
- Cat. No.:MDTC46-034-CI
- Storage temperature:Ambient
- Shipping temperature:Ambient