ContactUsLinkComponent
Ion selective electrode, ammonia
Ion selective electrode, ammonia
Catalog # 662-2809
Supplier:  Mettler - Toledo
undefined
Ion selective electrode, ammonia
Catalog # 662-2809
Supplier:  Mettler - Toledo
Specifications
  • Concentration:
    10-6 - 5×10⁻2 mol/l
  • Type:
    NH3 GSE
  • Description:
    Ion selective electrode, gas sensing, for ammonia (NH3)
  • Material:
    Plastic
  • Pk:
    1
  • Temp.:
    0...+50 °C
Specifications
About this item
This gas-sensing electrode is intended for reliably measuring the ammonia content (NH₃).
  • Modular and versatile
  • Refillable electrolyte
  • Polymer matrix membrane