Order Entry
Germany
ContactUsLinkComponent
Edelfosine ≥95% (by TLC), white solid, Millipore®
Edelfosine ≥95% (by TLC), white solid, Millipore®
Catalog # 341207-5
CAS Number:  
Edelfosine ≥95% (by TLC), white solid, Millipore®
Catalog # 341207-5
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :523.736 g/mol
Storage Temperature:Freezer
CAS Number:70641-51-9
Formula:C₂₇H₅₈NO₆P
MDL Number:MFCD00135190