Cation exchange chromatography columns, TSK-GEL® SCX
Supplier: TOSOH Bioscience
Certificates
About this item
TSK-Gel® SCX are polymer based stainless steel columns packed with porous polystyrene divinylbenzene polymer beads. Designed for the separation and analysis of organic acids, saccharides and alcohols.