Orders LinkContactUsLinkComponent
Extreme temperature cleanroom tapes
Extreme temperature cleanroom tapes
Supplier:   MICRONOVA
Extreme temperature cleanroom tapes
Supplier:  MICRONOVA
Certificates
Extreme temperature cleanroom tapes
Extreme temperature cleanroom tapes
4 Options Available Below
Product Details & Documents
These extreme temperature cleanroom tapes are made of polyamide film with silicone adhesive.
  • Flame resistant electrical insulation
  • Solder mask for circuit boards
  • Heat sealing and lamination
  • Cold storage
  • Lyophilising and freeze drying
Polyamide film with pressure-sensitive silicone adhesive. The extreme heat resistant properties (−73 to +260 °C) of this tape make it suitable for high heat and sub zero applications. The silicone adhesive ensures a clean release from most substrates even after being exposed to extreme temperatures.
Length: 32,9 mm
Colour: Amber transparent
Product Family Options
Product Information
  • Width
    Availability
    Price
  • 819-0067
    Cleanroom tape
    Cleanroom tape
    38,1 mm
    Discontinued
  • 817-0055
    Cleanroom tape
    Cleanroom tape
    6,3 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 28,10 €
    28,10 €
    1 * 1 Rolle
       28,10 €
  • 819-0071
    Cleanroom tape
    Cleanroom tape
    9,5 mm
    Discontinued
  • 817-0054
    Cleanroom tape
    Cleanroom tape
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 41,70 €
    41,70 €
    1 * 1 Rolle
       41,70 €
  • 819-0070
    Cleanroom tape
    Cleanroom tape
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 55,30 €
    55,30 €
    1 * 1 Rolle
       55,30 €
  • 817-0057
    Cleanroom tape
    Cleanroom tape
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 69,50 €
    69,50 €
    1 * 1 Rolle
       69,50 €
  • 819-0072
    Cleanroom tape
    Cleanroom tape
    76,2 mm
    Discontinued