Thioacetamide ≥99.0% ACS for the precipitation of heavy metals, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:62-55-5
:
Formula:CH₃CSNH₂
MDL Number:MFCD00008070
:75.13 g/mol
Melting Pt:108…114 °C
EINECS:200-541-4
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity (IR):complying
- Assay:99.0 - 101.0 %
- Melting range:111 - 114 °C
- Colour of the solution:complying
- Clearness of the solution:complying
- Residue on ignition (SO₄):Max. 0.05 %
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 100 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Application test:complying