Irradiated cleanroom tapes, CR100IR™
Supplier: MICRONOVA
Certificates
About this item
These ultra-processed irradiated cleanroom tapes are made of vinyl and are processed in a ISO 4 environment.
- Suitable for critical and aseptic processing applications, media preparation, batching, packaging within the laboratory or sterile processing area
- Helps to limit particulate and bioburden counts
- Extremely conformable; tapes and seals curved and irregular surfaces
- Clean peel from wafer cartridges and cleanroom work surfaces
- Matte finish can be written on with a cleanroom pen
- Wound on a 7.6 cm (3”) plastic core
The tape comes with full certification of irradiation and lot traceability.
Length: 33 m
Different colours available.
Packaging: Individually double bagged.
Specifications
- Catalog No:
- MICRCR100-2WHIR
- 819-0088
- MICRCR100-2TRIR
- MICRCR10034WHIR
- 819-0019
- 817-0067
- 819-0091
- MICRCR100-1LBIR
- MICRCR100-2REIR
- 819-0093
- Environmentally Preferable:
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient