Anti-GDPD5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-9207R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Glycerophosphodiester Phosphodiesterase Domain Containing 5
- Antigen symbol:GDPD5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:81544
- Antigen synonyms:GDE2|PP1665.|Gdpd5|GDPD5_HUMAN|Glycerophosphodiester phosphodiesterase domain-containing protein 5|Glycerophosphodiester phosphodiesterase domain containing 5|Glycerophosphodiester phosphodiesterase 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Promotes neurite formation. Cooperates with PRDX1 to drive postmitotic motor neuron differentiation. The glycerophosphodiester phosphodiesterase activity may be required for its role in neuronal differentiation. May contribute to the osmotic regulation of cellular glycerophosphocholine.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: GDPD5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat