Order Entry
Germany
ContactUsLinkComponent
Anti-NCL Rabbit Polyclonal Antibody (Cy7®)
Anti-NCL Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-8536R-CY7
Supplier:  Bioss
Anti-NCL Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-8536R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nucleolin
  • Antigen symbol:
    NCL
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    4691
  • Antigen synonyms:
    C23
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    631-662/710
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Nucleolin is the major nucleolar protein of growing eukaryotic cells. It is found associated with intranucleolar chromatin and pre-ribosomal particles. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly. May play a role in the process of transcriptional elongation. Binds RNA oligonucleotides with 5'-UUAGGG-3' repeats more tightly than the telomeric single-stranded DNA 5'-TTAGGG-3' repeats.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCL
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat