Anti-PROCA1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-9188R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Protein Interacting With Cyclin A1
- Antigen symbol:PROCA1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:147011
- Antigen synonyms:proline-rich cyclin A1-interacting protein|Protein interacting with cyclin A1|Protein PROCA1|RP23-185A18.6.|Proca1|PRCA1_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Belongs to the PROCA1 family.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PROCA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat