Order Entry
Germany
ContactUsLinkComponent
 
Anti-PROCA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-9188R-A647
 :  Bioss
Anti-PROCA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-9188R-A647
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein Interacting With Cyclin A1
  • Antigen symbol:
    PROCA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    147011
  • Antigen synonyms:
    proline-rich cyclin A1-interacting protein|Protein interacting with cyclin A1|Protein PROCA1|RP23-185A18.6.|Proca1|PRCA1_HUMAN
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Belongs to the PROCA1 family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PROCA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat