Specifications
- Antibody type:Primary
- Antigen name:Pyridoxal (pyridoxine vitamin B6) kinase
- Antigen symbol:PDXK
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8566
- Antigen synonyms:C21orf124|HEL-S-1a|PRED79|C21orf97|PNK|PKH
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Pyridoxal kinase belongs to the pyridoxine kinase family and phosphorylates vitamin B6, a step necessary for the conversion of vitamin B6 to pyridoxal 5’ phosphate (PLP), the active form of vitamin B6. PLP acts as a coenzyme and functions to maintain homeostasis. Pyridoxal kinase is a 312-amino acid cytoplasmic protein that may act as a homodimer and is expressed ubiquitously. There are three known isoforms of pyridoxal kinase, and isoform 3 expression is observed in adult testis and spermatozoa. The optimum pH for pyridoxal kinase is between 5.5 and 6.0. PDXK, the gene that encodes the pyridoxal kinase protein, maps to chromosome 21q22.3 and may be a candidate gene for autoimmune polyglandular disease type 1, a genetic disorder that has been mapped to the same region on chromosome 21.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDXK
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat