Anti-RGS4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-9452R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Regulator of G-protein Signaling 4
- Antigen symbol:RGS4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5999
- Antigen synonyms:RP11-430G6.1|MGC60244|MGC2124|SCZD9|RGP4|DKFZp761F1924
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Activity on G(z)-alpha is inhibited by phosphorylation of the G-protein. Activity on G(z)-alpha and G(i)-alpha-1 is inhibited by palmitoylation of the G-protein.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RGS4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat