Order Entry
Germany
ContactUsLinkComponent
Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-7949R-A350
Supplier:  Bioss
Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-7949R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amine Oxidase, Copper Containing 1
  • Antigen symbol:
    AOC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    26
  • Antigen synonyms:
    ABP1|DAO|KAO|DAO1|ABP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Catalyzes the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumor formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat