Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25, Member 28
- Antigen symbol:SLC25A28
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:81894
- Antigen synonyms:MFRN2|MRS3/4|NPD016|MRS4L
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Mitoferrin 2, also known as MRS3/4 (mitochondrial RNA-splicing protein 3/4 homolog), mitochondrial iron transporter 2, NPD016, MRS4L or SLC25A28 (solute carrier family 25 member 28), is a 364 amino acid multi-pass membrane protein of the mitochondrial inner membrane that mediates iron uptake. Mitoferrin 2 is thought to play a role in heme synthesis of hemoproteins and iron-sulfur cluster assembly. Ubiquitously expressed, Mitoferrin 2 is found at high levels in skeletal muscle, heart, placenta, kidney, lung, liver, brain and pancreas. Mitoferrin 2 is a member of the mitochondrial carrier family and undergoes alternative splicing events to produce four isoforms. Mitoferrin 2 contains three solcar repeats and is encoded by a gene that maps to human chromosome 10q24.2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC25A28
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat