Anti-DEFA6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-7129R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Defensin, alpha 6, Paneth cell-specific
- Antigen symbol:DEFA6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1671
- Antigen synonyms:HD-6|DEF6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/100
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DEFA6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat