Order Entry
Germany
ContactUsLinkComponent
Anti-DOCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-DOCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-7110R-A647
Supplier:  Bioss
Anti-DOCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-7110R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dedicator Of Cytokinesis 1
  • Antigen symbol:
    DOCK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1793
  • Antigen synonyms:
    DOCK180|ced5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. Functions as a guanine nucleotide exchange factor (GEF), which activates Rac Rho small GTPases by exchanging bound GDP for free GTP. Its GEF activity may be enhanced by ELMO1.Tissue specificity: Highly expressed in placenta, lung, kidney, pancreas and ovary. Expressed at intermediate level in thymus, testes and colon.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat