Anti-SCGB3A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6397R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Secretoglobin, Family 3A, Member 1
- Antigen symbol:SCGB3A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:92304
- Antigen synonyms:HIN-1|PnSP-2|UGRP2|LU105|HIN1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
SCGB3A1 is a potential growth inhibitory cytokine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCGB3A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human