Anti-RAB9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6177R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-9
- Antigen symbol:RAB9
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9367
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
RAB proteins are GTPases that regulate vesicular trafficking and reside in specific intracellular compartments. RAB9 has been localized to components of the endocytic/exocytic pathway. It has been implicated in the recycling of membrane receptors, such as the mannose 6-phosphate receptor from early endosomes to the trans Golgi network.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAB9
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat