Order Entry
Germany
ContactUsLinkComponent
Anti-SMARCB1 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-SMARCB1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-6109R-CY5.5
Supplier:  Bioss
Anti-SMARCB1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-6109R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SWI/SNF Related, Matrix Associated, Actin Dependent Regulator Of Chromatin, Subfamily B, Member 1
  • Antigen symbol:
    SMARCB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    6598
  • Antigen synonyms:
    hSNFS|SNF5|SNF5L1|Snr1|RTPS1|RDT|SWNTS1|Sfh1p|BAF47|INI1|MRD15|PPP1R144
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

SNF5 is involved in transcriptional activation. The SWI/SNF complex is required for the induced expression of a large number of genes. This complex alters chromatin structure to facilitate binding of gene-specific dedicated transcription factors. SNF5 also funtions as a tumor suppressor gene and is frequently mutated in malignant rhabdoid tumors. It is also involved in adipocyte differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMARCB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat