Anti-SMARCB1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6109R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SWI/SNF Related, Matrix Associated, Actin Dependent Regulator Of Chromatin, Subfamily B, Member 1
- Antigen symbol:SMARCB1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6598
- Antigen synonyms:hSNFS|SNF5|SNF5L1|Snr1|RTPS1|RDT|SWNTS1|Sfh1p|BAF47|INI1|MRD15|PPP1R144
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
SNF5 is involved in transcriptional activation. The SWI/SNF complex is required for the induced expression of a large number of genes. This complex alters chromatin structure to facilitate binding of gene-specific dedicated transcription factors. SNF5 also funtions as a tumor suppressor gene and is frequently mutated in malignant rhabdoid tumors. It is also involved in adipocyte differentiation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SMARCB1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat