Anti-DLL4 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6044R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Delta-like 4
- Antigen symbol:DLL4
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:54567
- Antigen synonyms:hdelta2|DLL4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:565-615/685
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in the Notch signaling pathway as Notch ligand. Activates NOTCH1 and NOTCH4. Involved in angiogenesis; negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. Essential for retinal progenitor proliferation is required for suppressing rod fates in late retinal progenitors as well as for proper generation of other retinal cell types. During spinal cord neurogenesis, inhibits V2a interneuron fate.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DLL4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat