- Antibody type:Primary
- Antigen name:RAD9 Checkpoint Clamp Component A
- Antigen symbol:RAD9
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:5883
- Antigen synonyms:RAD9 homolog|RAD 9 (S pombe) homolog|RAD9A|Cell cycle checkpoint control protein|RAD9 homolog A
- Modification:phospho-Ser380
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
This gene product is highly similar to Schizosaccharomyces pombe rad9, a cell cycle checkpoint protein required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein is found to possess 3' to 5' exonuclease activity, which may contribute to its role in sensing and repairing DNA damage. It forms a checkpoint protein complex with RAD1 and HUS1. This complex is recruited by checkpoint protein RAD17 to the sites of DNA damage, which is thought to be important for triggering the checkpoint-signaling cascade. Use of alternative polyA sites has been noted for this gene. [provided by RefSeq].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human