Order Entry
Germany
ContactUsLinkComponent
 
Anti-PRKD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BOSSBS-5563R-HRP
 :  Bioss
Anti-PRKD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BOSSBS-5563R-HRP
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase D1
  • Antigen symbol:
    PRKD1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5587
  • Antigen synonyms:
    PRKCM|Protein kinase D|nPKC-D1|PKD|Serine/threonine-protein kinase D1|PKD1|Protein kinase C mu type|PRKD1|nPKC-mu
  • Modification:
    phospho-Tyr463
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Members of the protein kinase C (PKC) family function in many extracellular receptor-mediated signal transduction pathways. See PRKCA (MIM 176960) for further background information. The PRKCM gene encodes a cytosolic serine-threonine kinase that binds to the trans-Golgi network and regulates the fission of transport carriers specifically destined to the cell surface.[supplied by OMIM]

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PRKD1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat