Order Entry
Germany
ContactUsLinkComponent
Anti-Wee1 Rabbit Polyclonal Antibody (Cy7®)
Anti-Wee1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5218R-CY7
Supplier:  Bioss
Anti-Wee1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5218R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Wee1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7465
  • Modification:
    phospho-Ser123
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    100-150/646
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Acts as a negative regulator of entry into mitosis (G2 to M transition) by protecting the nucleus from cytoplasmically activated cyclin B1-complexed CDK1 before the onset of mitosis by mediating phosphorylation of CDK1 on 'Tyr-15'. Specifically phosphorylates and inactivates cyclin B1-complexed CDK1 reaching a maximum during G2 phase and a minimum as cells enter M phase. Phosphorylation of cyclin B1-CDK1 occurs exclusively on 'Tyr-15' and phosphorylation of monomeric CDK1 does not occur. Its activity increases during S and G2 phases and decreases at M phase when it is hyperphosphorylated. A correlated decrease in protein level occurs at M/G1 phase, probably due to its degradation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Wee1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat