Anti-TAAR8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5060R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Trace Amine-associated Receptor 8
- Antigen symbol:TAAR8
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:83551
- Antigen synonyms:G-protein coupled receptor 102|GPR102|TRAR5.|Trace amine receptor 5|TAR5|TA5|TaR-5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
FADS1 is a component of a lipid metabolic pathway that catalyzes biosynthesis of highly unsaturated fatty acids from precursor essential polyunsaturated fatty acids, linoleic acid and alphanlinolenic acid. It catalyzes the desaturation of dihomo gamma linoleic acid and eicosatetraenoic acid to generate arachidonic acid and eicosapentaenoic acid respectively.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TAAR8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat