Anti-STAT2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-3428R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 2
- Antigen symbol:STAT2
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6773
- Antigen synonyms:IMD44|P113|ISGF-3|signal transducer and activator of transcription 2|STAT113
- Modification:phospho-Tyr690
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:670-720/851
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Signal transducer and activator of transcription that mediates signaling by type I IFNs (IFN-alpha and IFN-beta). Following type I IFN binding to cell surface receptors, Jak kinases (TYK2 and JAK1) are activated, leading to tyrosine phosphorylation of STAT1 and STAT2. The phosphorylated STATs dimerize, associate with IRF9/ISGF3G to form a complex termed ISGF3 transcription factor, that enters the nucleus. ISGF3 binds to the IFN stimulated response element (ISRE) to activate the transcription of interferon stimulated genes, which drive the cell in an antiviral state.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: STAT2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat