Order Entry
Germany
ContactUsLinkComponent
Anti-RGS5 Rabbit Polyclonal Antibody (Cy7®)
Anti-RGS5 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-2794R-CY7
Supplier:  Bioss
Anti-RGS5 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-2794R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Regulator of G-protein Signaling 5
  • Antigen symbol:
    RGS5
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    19737
  • Antigen synonyms:
    MSTP092|MSTP032|MSTP129|MST106|MSTP106|MST092|MST129
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    80-130/181
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G(i)-alpha and G(o)-alpha, but not to G(s)-alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat