Anti-HOXB4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-3650R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Homeobox B4
- Antigen symbol:HOXB4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3214
- Antigen synonyms:HOX-2.6|HOX2|HOX2F
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
HOXB4 is a homeodomain-containing transcription factor with diverse roles in embryonic development and the regulation of adult stem cells. It promotes proliferation of hematopoietic stem cells and can both activate and repress apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HOXB4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat