Anti-PLPP3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-4127R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Phospholipid Phosphatase 3
- Antigen symbol:PLPP3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8613
- Antigen synonyms:PAP2B|VCIP|PPAP2B|LPP3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:258-285/311
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Catalyzes the conversion of phosphatidic acid (PA) to diacylglycerol (DG). In addition it hydrolyzes lysophosphatidic acid (LPA), ceramide-1-phosphate (C-1-P) and sphingosine-1-phosphate (S-1-P). The relative catalytic efficiency is LPA = PA >C-1-P >S-1-P. May be involved in cell adhesion and in cell-cell interactions.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PLPP3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse