Order Entry
Germany
ContactUsLinkComponent
Anti-IL3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-IL3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2598R-A555
Supplier:  Bioss
Anti-IL3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2598R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 3
  • Antigen symbol:
    IL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    16187
  • Antigen synonyms:
    Multipotential colony-stimulating factor|P-cell-stimulating factor|MCGF|Hematopoietic growth factor|Mast-cell growth factor|Interleukin-3|IL3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    100-150/166
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Granulocyte/macrophage colony-stimulating factors are cytokines that act in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages. This CSF induces granulocytes, macrophages, mast cells, stem cells, erythroid cells, eosinophils and megakaryocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat