Order Entry
Germany
ContactUsLinkComponent
Anti-AMPD1 Rabbit Polyclonal Antibody (Cy5®)
Anti-AMPD1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-2077R-CY5
Supplier:  Bioss
Anti-AMPD1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-2077R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMP deaminase 1
  • Antigen symbol:
    AMPD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    229665
  • Antigen synonyms:
    Ampd1|Myoadenylate deaminase|Ampd-1|AMP deaminase isoform M|AI553520
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    125-175/745
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

AMP deaminase plays a critical role in energy metabolism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMPD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat